Order Entry
Austria
ContactUsLinkComponent
Horizontal racks, for multiwell plates, Nalgene®
Horizontal racks, for multiwell plates, Nalgene®
Catalog # NALG5039-0048
   Low Manufacturing Impact
undefined
Horizontal racks, for multiwell plates, Nalgene®
Catalog # NALG5039-0048
   Low Manufacturing Impact

Specifications

  • Shelves/drawers per rack:
    16 (4×4)
  • Description:
    Horizontal racks for multiwell plates
  • W×D×H:
    387×139×216 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1

Specifications

About this item

Stainless steel.

  • Racks fit 1,0 ml cryobank cryogenic storage vials
  • Horizontal design fits most refrigerators and freezers
  • Retainer clip on each compartment holds plates securely in place
  • Handles on tops and sides for easy transport
  • Label holder for easy identification of contents of rack

Securely store and transport samples in microplate, deep-well plates and cryobank racks using these horizontal storage racks.