TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
21123.
: Merck
:
- Pk:1
- Seq:0001
- Environmentally Preferable:
: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
:
:13,06793
:∼200 °C (1013 hPa)
:6 °C
:1.06 g/cm³ (20 °C)
:251 °C (closed cup)
:3082
:9,III
:GHS05
:GHS07
:GHS09
:Refrigerator
:9002-93-1
:9036-19-5
:MFCD00132505