Order Entry
Austria
ContactUsLinkComponent
N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane for GC derivatisation
N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane for GC derivatisation
Catalog # HYPETS-48915
CAS Number:  
N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane for GC derivatisation
Catalog # HYPETS-48915
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    10 Ampoul
  • Seq:
    0510

Specifications

About this item

 :
 :199.25 g/mol
EINECS:246-331-6
CAS Number:24589-78-4
Formula:C₆H₁₂F₃NOSi