Order Entry
Austria
ContactUsLinkComponent
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Catalog # HYPETS-38831
CAS Number:  
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Catalog # HYPETS-38831
CAS Number:  

Specifications

  • Pk:
    10 Ampoul
  • Seq:
    0510

Specifications

About this item

 :
 :257.40 g/mol
EINECS:247-103-9
 :GHS02
 :GHS05
CAS Number:25561-30-2
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269