Triethylammonium dihydrogentrifluoride
Catalog # MOLE11203893-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride
:
:161.21 g/mol
Density:1.094 g/cm³ (20 °C)
EINECS:277-550-5
UN:3265
ADR:8,II
CAS Number:73602-61-6
Formula:C₆H₁₈F₃N
MDL Number:MFCD00043294