ContactUsLinkComponent
Sodium dodecyl sulphate (SDS), LiChropur™ for ion pair chromatography, Supelco®
Sodium dodecyl sulphate (SDS), LiChropur™ for ion pair chromatography, Supelco®
  1.18309.0100
 :  Merck
 :  
undefined
Sodium dodecyl sulphate (SDS), LiChropur™ for ion pair chromatography, Supelco®
  1.18309.0100
 :  Merck
 :  
  • Pk:
    100 g
  • Seq:
    3401
 :
 :288.38 g/mol
 :14,08636
 :204…207 °C
 :1.1 g/cm³ (20 °C)
 :>100 °C
 :Ambient
 :205-788-1
 :1325
 :4.1,
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS05
 :GHS07
 :151-21-3
 :CH₃(CH₂)₁₁OSO₃Na
 :MFCD00036175