Order Entry
Austria
ContactUsLinkComponent
Acrylamide 40% in aqueous solution, Sigma-Aldrich®
Toxic
Health
Acrylamide 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIALA4058-100ML
Supplier:  Merck
CAS Number:  
undefined
Toxic
Health
Acrylamide 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIALA4058-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :71.08 g/mol
EINECS:201-173-7
UN:3426
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:79-06-1
Formula:C₃H₅NO
MDL Number:MFCD00008032