VWR® Acrylamide and Bis-Acrylamide
Supplier: VWR Chemicals
Certificates
About this item
An extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions and ready to use solutions for customised polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids.
Ultra-pure (>99,9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications.
Liquid blends minimise the handling of neurotoxic acrylamide.
Packaging: Packed in a plastic bottle.