Sodium triacetoxyborohydride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56553-60-7
:
Formula:C₆H₁₀BNaO₆
MDL Number:MFCD00012211
:211.94 g/mol
Melting Pt:114…118 °C (dec.)
UN:3396
ADR:4.3,II
:GHS05
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Assay:≥96.0 %
Specification Test Results
Frequently Bought Together