Order Entry
Austria
ContactUsLinkComponent
Acetonitrile with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Irritant
Health
Flammable
Acetonitrile with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # HONCLC441-2.5
CAS Number:  
Irritant
Health
Flammable
Acetonitrile with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # HONCLC441-2.5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, GL45
  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :75-05-8
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:–46 °C
Density:0.7767 g/cm³ (25 °C)
Flash Pt:8 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
 :GHS07
 :GHS08
 :GHS02