Lithium chloride 11,1% in aqueous solution
1
Catalog # HANNHI7111
Jump to:
Discontinued
Product Details & Documents
- CAS Number:7447-41-8
- Formula:LiCl
- MDL Number:MFCD00011078
- Molecular Weight:42.39 g/mol
- Storage Temperature:Ambient
- EINECS:231-212-3
Specifications
Specifications
- Pk:1



