Order Entry
Austria
ContactUsLinkComponent
 
Organic reference standard, Diethyl ether 2,000 µg/ml in methanol
 :  Restek
 
Organic reference standard, Diethyl ether 2,000 µg/ml in methanol
 :  Restek

 

  • CAS No.:
    60-29-7
  • Formula:
    C₄H₁₀O
  • Description:
    Diethyl ether

 

Frequently Bought Together