Cytochalasin B
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14930-96-2
:
Formula:C₂₉H₃₇NO₅
MDL Number:MFCD00077704
:479.62 g/mol
Melting Pt:218…223 °C
Storage Temperature:Refrigerator
EINECS:239-000-2
UN:1544
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:White to off-white Powder
- HPLC:≥98 %
Specification Test Results
Frequently Bought Together