Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
    
 
undefined
 
 
 
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

 

 

  • Autoignition Temperature:
    973 °F
  • Assay (GC):
    Min. 99.90 %
  • GC (as heptachlor epoxide):
    Max. 10 ng/l
  • Vapor Density:
    1.41 (vs air)
  • Non-volatile matter:
    Max. 0.0002 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Free alkali (as NH₃):
    Max. 0.0001 %
  • Absorbance at 400 nm:
    Max. 0.0044
  • Absorbance at 250 nm:
    Max. 0.0044
  • Absorbance at 235 nm:
    Max. 0.0044
  • Absorbance at 228 nm:
    Max. 0.005
  • Absorbance at 200 nm:
    Max. 0.02
  • Absorbance at 195 nm:
    Max. 0.10
  • Fluorescence (chinin) at 254 nm:
    Max. 0.5 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb
  • HPLC Gradient at 210 nm:
    Max. 1 mAU
  • HPLC Gradient at 254 nm:
    Max. 0.2 mAU
  • Baseline drift at 210 nm:
    Max. 12 mAU
  • Measuring Range:
    ≤12 mAU baseline drift, (210 nm)

 

 

Frequently Bought Together