Calcium carbonate, EMPROVE® api Ph. Eur., BP, USP suitable for use as an active pharmaceutical ingredient (max. 0.002% Fe), precipitated, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:471-34-1
:
Formula:CaCO₃
MDL Number:MFCD00010906
:100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C
Density:2.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
Frequently Bought Together