Bromelain (from Stem)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Specifications
- CAS No.:37189-34-7
- Source:Stem
- Storage conditions:Refrigerator
- Form:Cream to light brown powder
- Enzyme name:Bromelain
Specifications
Frequently Bought Together