Order Entry
Austria
ContactUsLinkComponent
Pesticide reference standard, 2,3,4,5,6-Pentachloroaniline 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, 2,3,4,5,6-Pentachloroaniline 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA15940000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 2,3,4,5,6-Pentachloroaniline 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA15940000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    527-20-8
  • Description:
    2,3,4,5,6-Pentachloroaniline 100 µg/ml in cyclohexane
  • Formula:
    C₆H₂Cl₅N
  • Description:
    2,3,4,5,6-Pentachloroaniline

Specifications

About this item