Order Entry
Austria
ContactUsLinkComponent
Dexamethasone
Dexamethasone
Catalog # PROOMM0213.00
CAS Number:  
Dexamethasone
Catalog # PROOMM0213.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :392.47 g/mol
Melting Pt:268…271 °C
EINECS:200-003-9
CAS Number:50-02-2
Formula:C₂₂H₂₉FO₅
MDL Number:MFCD00064136