

Dexamethasone, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:50-02-2
:
Formula:C₂₂H₂₉FO₅
MDL Number:MFCD00064136
:392.47 g/mol
Melting Pt:262…264 °C (lit.)
EINECS:200-003-9
:GHS07
:GHS08
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard C
- Loss on Drying:≤5,0%
- Optical Rotation:+72 ° to +80 ° (c=1, dioxane)
Specification Test Results
Frequently Bought Together