Chrysin 97%, Sigma-Aldrich®
Catalog # C80105-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance (Colour):Faint yellow to yellow and faint beige to beige and faint brown to light brown
- Appearance (Form):Powder
- Titration (Nt) Tbaoh 0.1M:96.5 - 103.5 %
- Purity (HPLC Area %):≥96.5 %
- Solubility (Colour):Yellow to Amber
- Solubility (Turbidity):Clear to slightly hazy
- Solubility (Method):50Mg/Ml, Pyridin
- Infrared Spectrum:Conforms to Structure
- Wavelength (1) (Uv):344 - 352 Nm
- Molar Absorbancy Index (1):≥8000
- Wavelength (2) (Uv):278 - 286 Nm
- Molar Absorbancy Index (2):≥22000
- Wavelength (3) (Uv):259 - 267 Nm
- Molar Absorbancy Index (3):≥20000
- Wavelength (4) (Uv):220 - 228 Nm
- Molar Absorbancy Index (4):≥27000
- Uv Method:In 0.1N NaOH
Specification Test Results
About this item
:
:254.24 g/mol
Boiling Pt:429 °C (1013 hPa)
Melting Pt:284…286 °C (lit.)
:M-Clarity™ quality level = MQ100
Storage Temperature:Room Temperature
EINECS:207-549-7
CAS Number:480-40-0
Formula:C₁₅H₁₀O₄
MDL Number:MFCD00006834