
3-Bromo-2,6-difluorobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:398456-82-1
:
Formula:C₇H₃BrF₂O
MDL Number:MFCD08282773
:221 g/mol
Melting Pt:53…57 °C
EINECS:None
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together