3,5-Dichloro-4-hydroxybenzaldehyde
Lieferant: CHEMPUR
Zertifikate
Über diesen Artikel
CAS-Nummer:2314-36-5
:
Formel:C₇H₄Cl₂O₂
MDL-Nummer:MFCD00017605
:191.01 g/mol
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Lieferant: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Lieferant: VWR Chemicals
VWR® Facial Tissues
Lieferant: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
VWR®, Nitrile Gloves
Lieferant: VWR Collection
pH indicator strips, MQuant®, Supelco®
Lieferant: Merck
Sealing film for manual application, Parafilm® M
Lieferant: HEATHROW
Acetone ≥99%, TECHNICAL
Lieferant: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Lieferant: VWR Collection
VWR®, Transfer Pipettes, High Performance
Lieferant: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Lieferant: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Lieferant: VWR Collection