Order Entry
Austria
ContactUsLinkComponent
2-Nitro-4-(trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®
2-Nitro-4-(trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL324779-5G
Supplier:  Merck
CAS Number:  
2-Nitro-4-(trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL324779-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :289.62
Boiling Pt:325 °C (1013 hPa)
Melting Pt:42…44 °C
ADR:8,II
 :M-Clarity™ quality level = MQ100
UN:3261
CAS Number:837-95-6
Formula:C₇H₃ClF₃NO₄S
MDL Number:MFCD00013456