2-Mercapto-5-methoxybenzimidazole ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37052-78-1
:
Formula:C₈H₈N₂OS
MDL Number:MFCD00134581
:180.23 g/mol
Boiling Pt:400 °C (1013 hPa)
Melting Pt:255…259 °C
Storage Temperature:Ambient
EINECS:253-326-2
:GHS07
Related Information
Frequently Bought Together