Order Entry
Austria
ContactUsLinkComponent
2-Mercapto-5-methoxybenzimidazole
2-Mercapto-5-methoxybenzimidazole
Catalog # MOLE66787209-25G
Supplier:  Molekula
CAS Number:  
2-Mercapto-5-methoxybenzimidazole
Catalog # MOLE66787209-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :180.23 g/mol
Boiling Pt:400 °C (1013 hPa)
Melting Pt:261…263 °C
EINECS:253-326-2
CAS Number:37052-78-1
Formula:C₈H₈N₂OS
MDL Number:MFCD00134581