2-Mercapto-5-methoxybenzimidazole 99+%
:37052-78-1
:
:C₈H₈N₂OS
:MFCD00134581
:180.23 g/mol
:400 °C (1013 hPa)
:255…262 °C
:253-326-2
:GHS07
- Appearance:White to yellow to light beige Powder
- Infrared spectrum:Conforms
- Melting point:255 to 260 °C
- HPLC:≥99.0 %
Frequently Bought Together