

2-Ethylhexane-1,3-diol (mixture of isomers) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:94-96-2
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00004578
:146.23 g/mol
Boiling Pt:241…249 °C
Melting Pt:–40 °C
Density:0.9422 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-377-9
:GHS05
:GHS07
Related Information
Frequently Bought Together