Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-100.5% Ph. Eur., USP
:77-86-1
:
:NH₂C(CH₂OH)₃
:MFCD00004679
:121.14 g/mol
:13,09842
:357 °C (1013 hPa)
:169…172 °C
:1.328 g/cm³ (20 °C)
:201-064-4
:GHS07
:Ambient
- Appearance:Conforms (see CoA/CoS)
- Heavy metals (as Pb):Max. 10 ppm
- Residual solvents Class 2 (ICH Q3C) ::Passes test USP
- Methanol:Below option 1 limit
- Other residual solvents(Ph.Eur./ICH):Unlikely by manuf.process
- Residues of metal catalysts or reagents:Unlikely by manuf.process
- As (Arsenic):Max. 2 ppm
- Cd (Cadmium):Max. 1 ppm
- Co (Cobalt):Max. 1 ppm
- Hg (Mercury):Max. 2 ppm
- Ni (Nickel):Max. 50 ppm
- Pb (Lead):Max. 3 ppm
- V (Vanadium):Max. 2 ppm
- Conforms to Ph.Eur./USP:Passes test
- Ph.Eur. specifications::Max. 0.1 %
- Assay (calculated on dried) [Ph.Eur.]:99.0 --> 100.5 %
- Identification B [Ph.Eur.]:168 --> 174 °C
- Identification C:Passes test Ph.Eur.
- Appearance of solution:Passes test Ph.Eur.
- pH (20°C; 5 %) [Ph.Eur.]:10.0 --> 11.5
- Related substances [Ph.Eur.]:Max. 1.0 %
- Cl (Chloride) [Ph.Eur.]:Max. 100 ppm
- Fe (Iron) [Ph.Eur.]:Max. 10 ppm
- Loss on drying (105°C) [Ph.Eur.]:Max. 0.5 %
- Sulphated ash [Ph.Eur.]:Max. 0.1 %
- Assay (calculated on dried basis) [USP]:99.0 --> 101.0 %
- Identification A:Passes test USP
- Identification B:Passes test USP
- Residue on ignition [USP]:Max. 0.1 %
- Melting range or temperature [USP]:168 --> 172 °C
- pH (5 %) [USP]:10.0 --> 11.5
- Loss on drying (105°C; 3 h) [USP]:Max. 1.0 %
Frequently Bought Together