Order Entry
Austria
ContactUsLinkComponent
1-Propanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
1-Propanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates
undefined
1-Propanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Free acid (as C₂H₅COOH):
    Max. 0.001 %
  • Transmittance at 220 nm:
    Min. 40 %
  • Transmittance at 240 nm:
    Min. 85 %
  • Transmittance from 275 nm:
    Min. 99 %
  • Vapor Pressure:
    14.9 mmHg ( 20 °C)
  • Autoignition Temperature:
    700 °F
  • Vapor Density:
    2.1 (vs air)
  • Refractive Index:
    n20/D 1.384 (lit.)

Specification Test Results

Related Information

Frequently Bought Together