Order Entry
Austria
ContactUsLinkComponent
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Flammable
Irritant
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.9 %
  • Acidity:
    Max. 0.0003 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Absorbance (200 nm):
    Max. 0.1
  • Absorbance (210 nm):
    Max. 0.1
  • Absorbance (220 nm):
    Max. 0.05
  • Absorbance (230 nm):
    Max. 0.01
  • Absorbance (250 nm):
    Max. 0.005
  • Transmittance (200 nm):
    Min. 80 %
  • Transmittance (210 nm):
    Min. 85 %
  • Transmittance (220 nm):
    Min. 90 %
  • Transmittance (230 nm):
    Min. 98 %
  • Transmittance (250 nm):
    Min. 98 %

Specification Test Results

Frequently Bought Together