Order Entry
Austria
ContactUsLinkComponent
2-Nitro-1-naphthol
2-Nitro-1-naphthol
Catalog # MOLE23574535-5G
Supplier:  Molekula
CAS Number:  
2-Nitro-1-naphthol
Catalog # MOLE23574535-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1-Hydroxy-2-nitronaphthalene
 :
 :189.17 g/mol
Melting Pt:123…125 °C
EINECS:210-131-7
CAS Number:607-24-9
Formula:C₁₀H₇NO₃
MDL Number:MFCD00003956