Pesticide reference standard, 2,3,5,6-Tetrachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17375600CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:935-95-5
- Description:2,3,5,6-Tetrachlorophenol 10 µg/ml in cyclohexane
- Formula:C₆H₂Cl₄O
- Description:2,3,5,6-Tetrachlorophenol