Order Entry
Austria
ContactUsLinkComponent
Pesticide reference standard, 2,3,4,6-Tetrachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, 2,3,4,6-Tetrachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL17374600CY
 :  
Pesticide reference standard, 2,3,4,6-Tetrachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL17374600CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    58-90-2
  • Description:
    2,3,4,6-Tetrachlorophenol 10 µg/ml in cyclohexane
  • Formula:
    C₆H₂Cl₄O
  • Description:
    2,3,4,6-Tetrachlorophenol