Pesticide reference standard, 3-Chlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11471000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:108-43-0
- Description:3-Chlorophenol 100 µg/ml in methanol
- Formula:C₆H₅ClO
- Description:3-Chlorophenol