Spermidine trihydrochloride
Catalog # EHERC16972738
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:254.63 g/mol
Merck Index:12,08893
Melting Pt:256…258 °C
Storage Temperature:Freezer
EINECS:206-379-0
CAS Number:334-50-9
Formula:C₇H₂₂Cl₃N₃
MDL Number:MFCD00012918