Pesticide reference standard, 1,4-Dichloronaphthalene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20421400AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:1825-31-6
- Description:1,4-Dichloronaphthalene 10 µg/ml in acetonitrile
- Formula:C₁₀H₆Cl₂
- Description:1,4-Dichloronaphthalene