Pesticide reference standard, 3,4-Dichlorophenol 10 µg/ml in methanol, Dr Ehrenstorfer
EHERL12453400ME
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:95-77-2
- Description:3,4-Dichlorophenol 10 µg/ml in methanol
- Formula:C₆H₄Cl₂O
- Description:3,4-Dichlorophenol