Triethylamine
Catalog # PIER25108
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: (Diethylamino)ethane, TEA
:
:101.19 g/mol
Flash Pt:–11 °C
EINECS:204-469-4
UN:1296
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:121-44-8
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051
Boiling Pt:90
Melting Pt:–115 °C
Density:0.729 g/cm³ (20 °C)
ADR:3 II
Merck Index:13,09740