Orders LinkContactUsLinkComponent
Laboratory papers, Nalgene®
Polyolefin plastic paper sheets, 216×279 mm
  NALG6304-9811
   Low Manufacturing Impact
Laboratory papers, Nalgene®
  NALG6304-9811
   Low Manufacturing Impact
Laboratory papers, Nalgene®
Laboratory papers, Nalgene®
Polyolefin plastic paper sheets, 216×279 mm
126,00 €
1 * 50 ST
These polyolefin sheets roll or fold without cracking and stay flexible at cold temperatures. They won't rip, stretch or shrink. They can be punched for use in binders or cut into labels. These sheets do not accept pencil.
  • Resistant to many solvents including acetone, formaldehyde, formalin, xylene, acetic acid and ethanol
  • Accept many inks including laser, inkjet, copier, markers, ballpoint, gels and technical markers
Format: A4
  • Description:
    Polyolefin plastic paper sheets, 216×279 mm
  • Cat. No.:
    NALG6304-9811
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact