


Acetonitrile ≥99.9% (by GC), CHROMASOLV™ NMR for LC-NMR, Riedel-de Haën™
34955-1L
:
- Pack type:Glass bottle, narrow mouth
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Autoignition Temperature:973 °F
- Assay (GC):Min. 99.9 %
- Non-volatile matter:Max. 0.0002 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Free alkali (as NH₃):Max. 0.0001 %
- Suitability for the LC-NMR:complying
- Propionitrile (GC):Max. 0.003 %
- Identity (NMR):complying
- Vapor Density:1.41 (vs air)
- Explosion Limit:16 %
:
:41.05 g/mol
:13,00071
:82 °C (1.013 hPa)
:–46 °C
:0.7857 g/cm³ (20 °C)
:6 °C
:Ambient
:200-835-2
:1648
:3,II
:01-2119471307-38
:GHS08
:GHS07
:GHS02
:75-05-8
:H₃CCN
:MFCD00001878