


Acetonitrile ≥99.9% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™
:75-05-8
:
:H₃CCN
:MFCD00001878
:41.05 g/mol
:13,00071
:82 °C (1.013 hPa)
:–46 °C
:0.7857 g/cm³ (20 °C)
:6 °C
:Ambient
:200-835-2
:1648
:3,II
:01-2119471307-38
:GHS08
:GHS07
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Vapor Density:1.41 (vs air)
- Assay (GC):Min. 99.9 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.02 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Free alkali (as NH₃):Max. 0.0001 %
- Transmittance at 195 nm:Min. 75 %
- Transmittance at 200 nm:Min. 93 %
- Transmittance from 230 nm:Min. 99 %
- Baseline drift at 210 nm:Max. 15 mAU
- HPLC Gradient at 210 nm:Max. 3 mAU
- Fluorescence (chinin) at 254 nm:Max. 1 ppb
- Fluorescence (chinin) at 365 nm:Max. 1 ppb
- for AMD technic:complying
- Vapor Pressure:72.8 mmHg ( 20 °C)
- Explosion Limit:16 %
- Refractive Index:n20/D 1.344 (lit.)
Frequently Bought Together