Orders LinkContactUsLinkComponent
Dropping pipettes
Dropping pipettes
Supplier:   Brand
Dropping pipettes
Supplier:  Brand
Certificates
Dropping pipettes
Dropping pipettes
2 Options Available Below
Product Details & Documents
LDPE, graduated. For sampling and decanting infectious or toxic liquids.
Product Family Options
Product Information
  • Capacity
    Length
    Packed
    Availability
    Price
  • 1,5 ml
    133 mm
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 27,70 €
    27,70 €
    1 * 100 ST
       27,70 €
  • 5 ml
    194 mm
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 30,80 €
    30,80 €
    1 * 100 ST
       30,80 €