Order Entry
Austria
Orders LinkContactUsLinkComponent
Dropping pipettes

Dropping pipettes

1,8 ml, 98 mm, 250/pack
Catalog # BRND125400
Supplier:   Brand
Dropping pipettes
Catalog # BRND125400
Supplier:  Brand
Dropping pipettes
Dropping pipettes

1,8 ml, 98 mm, 250/pack

Price
111,00 €
Log in for availability
1 * 250 ST
Product Details & Documents

LDPE. Dropping pipette with integrated pipetting bulb.

Specifications
  • Packed:
    250/pack
  • Capacity:
    1,8 ml
  • Length:
    98 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.