ContactUsLinkComponent
Hydrometer cylinder, Nalgene®
Hydrometer cylinder, Nalgene®
  NALG6230-0500
   Low Manufacturing Impact
undefined
Hydrometer cylinder, Nalgene®
  NALG6230-0500
   Low Manufacturing Impact
  • Capacity:
    500 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Pk:
    1
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Made of PMP, clear.
  • Superior chemical resistance
  • With broad base
  • Good temperature resistance
PMP protect against breakage with the hydrometer cylinder. Lighter in weight than glass. This transparent, autoclavable cylinder can also be used as a lactometer cylinder.