Orders LinkContactUsLinkComponent
Pasteur pipettes, disposable
Pasteur pipettes, disposable
Supplier:   Brand
Pasteur pipettes, disposable
Supplier:  Brand
Certificates
Pasteur pipettes, disposable
Pasteur pipettes, disposable
6 Options Available Below
Product Details & Documents
LDPE.
  • Excellent at reproducing number of drops per mililitre
  • Ideal for aliquots
  • Pasteur pipettes can be filled and deep frozen, or changed into a closed vessel by heat-sealing the tip
  • The integrated pipetting bulb depresses easily, minimising fatigue from frequent pipetting
  • Resistant to gas or gamma radiation sterilisation
Product Family Options
Product Information
  • Capacity
    Drops per ml
    Stem Ø
    Length
    Graduation
    Sterile
    Packed
    Availability
    Price
  • 1,0 ml
    70
    1,0 mm
    144 mm
    0,25 ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 111,00 €
    111,00 €
    1 * 500 ST
       111,00 €
  • 2,0 ml
    22 - 26
    3,3 mm
    152 mm
    2/0,5 ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 60,20 €
    60,20 €
    1 * 500 ST
       60,20 €
  • 3,0 ml
    25 - 27
    2,8 mm
    152 mm
    - ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 53,50 €
    53,50 €
    1 * 500 ST
       53,50 €
  • 3,5 ml
    25 - 30
    3,4 mm
    151 mm
    1/0,25 ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 44,10 €
    44,10 €
    1 * 500 ST
       44,10 €
  • 3,5 ml
    21 - 28
    3,2 mm
    152 mm
    3/0,5 ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 43,10 €
    43,10 €
    1 * 500 ST
       43,10 €
  • 4,0 ml
    25
    3,0 mm
    150 mm
    - ml
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 109,00 €
    109,00 €
    1 * 500 ST
       109,00 €