Order Entry
Austria
ContactUsLinkComponent
Acetonitrile ≥99.8% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Flammable
Irritant
Acetonitrile ≥99.8% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # 9255-02
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Acetonitrile ≥99.8% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # 9255-02
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 5
  • FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤5
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤10
  • Assay (CH₃CN) (by GC, corrected for water):
    ≥99.8 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤1.0 ppm
  • Titrable Acid (µeq/g):
    ≤0.3
  • Titrable Base (µeq/g):
    ≤0.6
  • Water (by KF, coulometric):
    ≤0.05 %

Specification Test Results

About this item

 :
 :41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
 :Packaged under inert gas.
REACH:01-2119471307-38
 :GHS02
 :GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878