Acetonitrile, anhydrous ≥99.8%
:75-05-8
:
:H₃CCN
:MFCD00001878
:41.05 g/mol
:14,70
:80…82 °C
:–48…–45 °C
:0.786
:5 °C (41 °F)
:Ambient
:200-835-2
:1648
:3,II
:01-2119471307-38
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Frequently Bought Together