Desiccators, with knob lid
Supplier: Brand
Certificates
About this item
Borosilicate 3.3 glass with precisely ground flat flanges.
- Base ring surface is studded to improve stability and help prevent cracks
- Interchangeable knobbed lid
Certifications: DIN 12491
Frequently Bought Together