Order Entry
Austria
Orders LinkContactUsLinkComponent
Hexanal

Hexanal

Glass bottle, 100 mL
Catalog # ROTH3327.1
Flammable
Irritant
Marketsource
Supplier:   Roth Carl
Hexanal
Catalog # ROTH3327.1
Supplier:  Roth Carl
Marketsource
CAS Number:  66-25-1
Hexanal

Glass bottle, 100 mL

CAS Number:  66-25-1
Price
49,30 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Hexylaldehyde, Caproaldehyde, Hexaldehyde, Aldehyde C6, Hexyl aldehyde
  • CAS Number:66-25-1
  • Formula:C₆H₁₂O
  • MDL Number:MFCD00007027
  • Molecular Weight:100.16 g/mol
  • Merck Index:13,01766
  • Boiling Pt:131 °C (1013 hPa)
  • Melting Pt:–56 °C
  • Density:0.81 g/cm³ (20 °C)
  • Flash Pt:25 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-624-5
  • UN:1207
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications