Order Entry
Austria
ContactUsLinkComponent
1,2-Dibromo-3-chloropropane
Toxic
Health
1,2-Dibromo-3-chloropropane
Catalog # EHERCA12235000
Supplier:  EHRENSTORFER
CAS Number:  
Toxic
Health
1,2-Dibromo-3-chloropropane
Catalog # EHERCA12235000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
Merck Index:12,03068
Boiling Pt:195 °C (1013 hPa)
Melting Pt:6.1 °C
Density:2.085 g/cm³ (9 °C)
Storage Temperature:Ambient
EINECS:202-479-3
UN:2872
ADR:6.1,III
 :236.33 g/mol
 :GHS06
 :GHS08
CAS Number:96-12-8
Formula:ClH₂CBrCHCH₂Br
MDL Number:MFCD00039365